accessibility menu, dialog, popup

UserName

NH2NNH2HOONH2HOOH2NOHOOHO

CAS RN 991-01-5

IUPAC Name: 3,5-bis(3-amino-3-carboxypropyl)-2-(4-amino-4-carboxybutyl)-1-(5-amino-5-carboxypentyl)pyridin-1-ium
Synonyms: isodesmosine iso-desmosin
Molecular Weight (g/mol): 526.61
Molecular Formula: C24H40N5O8
InChi Key: RGXCTRIQQODGIZ-UHFFFAOYNA-O
SMILES: NC(CCCC[N+]1=CC(CCC(N)C(O)=O)=CC(CCC(N)C(O)=O)=C1CCCC(N)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results