missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Paraben, TRC

Product Code. 30010345
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010345 10 g 10g
30008807 100 g 100g
30004324 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010345 Supplier Toronto Research Chemicals Supplier No. TRCI78010010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4247-02-3
Molecular Formula C11 H14 O3
Molecular Weight (g/mol) 194.23
Synonym 2-Methylpropyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate Imp. E (EP), Isobutyl Parahydroxybenzoate
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
SMILES CC(C)COC(=O)c1ccc(O)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H14O3/c1-8(2)7-14-11(13)9-3-5-10(12)6-4-9/h3-6,8,12H,7H2,1-2H3
Formula Weight 194.0943
Percent Purity >95%
Chemical Name or Material Isobutyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.