accessibility menu, dialog, popup

UserName

CAS RN 4247-02-3

H3CCH3OOOH

CAS RN 4247-02-3

IUPAC Name: 2-methylpropyl 4-hydroxybenzoate
Synonyms: isobutylparaben
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: XPJVKCRENWUEJH-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results