Learn More
Iron Dextran, TRC
Product Code. 30133035
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133035 100 g 100g
30004696 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30133035 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCI774950100G
 Restricted Item
Estimated Shipment: 27-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 9004-66-4
Molecular Formula (C6H10O5)n[Fe(OH)3]m
Molecular Weight (g/mol) 153.93
Synonym Imferon, A 100, B 75, Chinofer, CosmoFer, Dexfer, Dexferrum, Dextran Iron Complex, Dextran Magnetite, Dextran-iron Oxide, Dextrofer 100, Dextrofer 75, Fe-dextran, Fenate, Ferdex 100, Ferran 100, Ferranimal 75, Ferranimal 75M, Ferric Dextran, Ferridextran, Ferrodex 100, Ferrodextran, Ferroglucin, Ferroglukin, Ferroglukin 75, Hyferdex, INFeD, Idofer, Impheron, Imposil, Iro-jex, Iron Dextran Injection, Iron-GARD, Iron-dextran Complex, Ironorm, Kemofei, Myofer 100, Nonemic, Polyfer, Proferedex, Prolongal, SHU 555;
SMILES OS(=O)(=O)O.[Fe]
Spezifikation
Quantity 100 g
InChI Formula InChI=1S/Fe.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)
Chemical Name or Material Iron Dextran