accessibility menu, dialog, popup

UserName

CAS RN 9004-66-4

OHSOOHOFe

CAS RN 9004-66-4

IUPAC Name: sulfuric acid iron
Synonyms: copperas ferrous sulfate ferrous sulphate
Molecular Weight (g/mol): 153.92
Molecular Formula: FeH2O4S
InChi Key: MVZXTUSAYBWAAM-UHFFFAOYSA-N
SMILES: [Fe].OS(O)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results