Learn More
Ipronidazole, TRC
Product Code. 17914005
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914005 50 mg 50mg
17990345 100 mg 100mg
17921615 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17914005 Supplier Toronto Research Chemicals Supplier No. TRCI74700050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14885-29-1
Molecular Formula C7 H11 N3 O2
Molecular Weight (g/mol) 169.18
Synonym Imidazole, 2-isopropyl-1-methyl-5-nitro- (8CI), 1-Methyl-2-(1-methylethyl)-5-nitro-1H-imidazole, 1-Methyl-2-isopropyl-5-nitroimidazole, 2-Isopropyl-1-methyl-5-nitroimidazole, Ipronidazole, Ipropran, NSC 109212, Ro 07-1554
IUPAC Name 1-methyl-5-nitro-2-propan-2-ylimidazole
SMILES CC(C)c1ncc([N+](=O)[O-])n1C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3
Formula Weight 169.0851
Percent Purity >95%
Chemical Name or Material Ipronidazole
Grade HPLC