Learn More
Invitrogen™ BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit
Product Code. 10798453
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10798453 20 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10798453 Supplier Invitrogen™ Supplier No. V48820
 Restricted Item
Estimated Shipment: 04-12-2026
Includes
The BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit contains the pLenti4/BLOCK-iT™-DEST vector, pLenti4-GW/H1/TO-lamin shRNA control vector, and One Shot™ Stbl3™ Chemically Competent E.Coli.
For lentiviral delivery and genomic integration of DNA coding for shRNA
The BLOCK-iT Lentiviral RNAi Zeo Gateway Vector Kit contains the pLenti4/BLOCK-iT-DEST expression vector which enables lentiviral delivery and genomic integration of DNA coding for shRNA. Once expressed, the shRNA is processed by cellular machinery and initiates target-specific RNAi.
  • Gateway Technology for efficient recombination of the RNAi cassette from the BLOCK-iT inducible pENTR™/H1/TO vector
  • All required components for efficient lentiviral packaging, delivery, and integration of the shRNA
  • Zeocin selection marker for fast selection of clonal cell lines containing the RNAi cassette
Lentiviral shRNA and miRNA, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Virus-Based RNAi
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNAi Expression Vector Kit
Content And Storage The BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit contains the pLenti4/BLOCK-iT™-DEST vector, pLenti4-GW/H1/TO-lamin shRNA control vector, and One Shot™ Stbl3™ Chemically Competent E.Coli. Store the pLenti4/BLOCK-iT™-DEST, pLenti6/TR vector, Blasticidin, ViraPower™ Lentiviral packaging mix, and the control vector at -20°C. Store the One Shot Stbl3™ Competent Cells 293 FT cell, and LR Clonase™ enzyme mix at -80°C. Store Lipofectamine™ 2000 at +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Cloning Method Gateway™
Constitutive or Inducible System Inducible
Delivery Type Lentiviral
RNAi Type shRNA
Selection Agent (Eukaryotic) Zeocin™
Vector pLenti
Promoter H1/TO
Format Kit
Product Line BLOCK-iT, Gateway
Quantity 20 Reactions
Afficher plus Afficher moins
For Research Use Only. Not for use in diagnostic procedures.
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.