missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 2/CD18 Antibody (MEM-48), FITC, Novus Biologicals™

Product Code. 18214570 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214570 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214570 Supplier Novus Biologicals Supplier No. NB500380
  • ONLINE EXCLUSIVE PRICE
    559.00 EUR
    528.15 EUR / 100 tests
    Save 30.85 EUR
    5% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Canine (Negative), Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 2/CD18
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-48
Conjugate FITC
Dilution Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Gene Symbols ITGB2
Host Species Mouse
Immunogen Leukocytes of a patient suffering from a LGL-type leukemia.
Molecular Weight of Antigen 95 kDa
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3689
Test Specificity The antibody MEM-48 recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta2 subunit; beta2 integrin). CD18 is a 90-95 kDa type I transmembrane protein expressed on all leukocytes.
Target Species Canine (Negative), Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.