missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (MEM-148), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Porcine (Negative) samples. It is validated for CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot.
Specifications
Specifications
| Antigen | Integrin beta 2/CD18 |
| Applications | CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MEM-148 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Accession No. | P05107 |
| Gene Alias | 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 |
| Gene Symbols | ITGB2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?