Learn More
MedChemExpress Ingenol
Product Code. 16434907
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16434907 Supplier MedChemExpress Supplier No. HYN0865
 Restricted Item
Estimated Shipment: 06-11-2026
MedChemExpress Ingenol is a PKC activator, with a Ki of 30 μM, with antitumor activity.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 348.43 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 30220-46-3
Product Type Natural Products
Purity 98.17%
Solubility DMSO : 100 mg/mL (287.00 mM∣Need ultrasonic)
Synonym (-)-Ingenol
Target PKC
Quantity 50 mg