missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-thiocarboxamide, 97%, Thermo Scientific™

Product Code. 15420738
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420738 1 g 1g
15430738 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420738 Supplier Thermo Scientific Alfa Aesar Supplier No. H54438.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59108-90-6
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.237
MDL Number MFCD01993552
InChI Key PPWNZMJWESSLGE-UHFFFAOYSA-N
Synonym 1h-indole-3-carbothioic acid amide, indole-3-thiocarboxamide, 9ci
PubChem CID 12276962
IUPAC Name 1H-indole-3-carbothioamide
SMILES C1=CC=C2C(=C1)C(=CN2)C(=S)N

Specifications

Quantity 1 g
Formula Weight 176.24
Percent Purity 97%
Chemical Name or Material Indole-3-thiocarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.