accessibility menu, dialog, popup

UserName

HNSNH2

CAS RN 59108-90-6

IUPAC Name: 1H-indole-3-carbothioamide
Synonyms: 1H-indole-3-carbothioamide
Molecular Weight (g/mol): 176.24
Molecular Formula: C9H8N2S
InChi Key: PPWNZMJWESSLGE-UHFFFAOYSA-N
SMILES: NC(=S)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results