Learn More
Indole-3-acetonitrile, TRC
Product Code. 17993035
Change view
Click to view available options
Quantity:
250 mg
5 g
10 g
Unit Size:
10g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993035 5 g 5g
17935035 10 g 10g
17950605 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993035 Supplier Toronto Research Chemicals Supplier No. TRCI5773555G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 771-51-7
Molecular Formula C10 H8 N2
Molecular Weight (g/mol) 156.18
Synonym Indole-3-acetonitrile, 3-Indoleacetonitrile
IUPAC Name 2-(1H-indol-3-yl)acetonitrile
SMILES N#CCc1c[nH]c2ccccc12
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H8N2/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5H2
Formula Weight 156.0687
Chemical Name or Material 3-Indolyl-acetonitrile