accessibility menu, dialog, popup

UserName

NHN

CAS RN 771-51-7

IUPAC Name: 2-(1H-indol-3-yl)acetonitrile
Synonyms: indole-3-acetonitrile IAN usaf CB-29 usaf cb-29
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: DMCPFOBLJMLSNX-UHFFFAOYSA-N
SMILES: N#CCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)