missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-11R alpha Polyclonal specifically detects IL-11R alpha in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | IL-11R alpha |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q14626 |
| Gene Alias | IL-11 receptor subunit alpha, IL-11R subunit alpha, IL-11RA, IL-11R-alpha, interleukin 11 receptor, alpha, interleukin-11 receptor alpha chain, interleukin-11 receptor subunit alpha, MGC2146 |
| Gene Symbols | IL11RA |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?