missing translation for 'onlineSavingsMsg'
Learn More

IgG3 Lambda Human, APC, Cy7, Isotype Control, Novus Biologicals™

Product Code. 18363923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363923 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363923 Supplier Novus Biologicals Supplier No. NBP306871APCCY7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Polyclonal Isotype Control

IgG3 Lambda Polyclonal antibody specifically detects IgG3 Lambda in Human samples. It is validated for Control
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG3 Lambda
Classification Polyclonal
Conjugate APC-Cyanine7
Formulation PBS
Host Species Human
Immunogen Human myeloma serum
Quantity 0.1 mL
Regulatory Status RUO
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Isotype Controls
Isotype IgG3 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.