missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-3 Antibody (RM0096-11B16), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18214685 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214685 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214685 Supplier Novus Biologicals Supplier No. NBP122452AF647
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody

IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-3
Applications Western Blot
Classification Monoclonal
Clone RM0096-11B16
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Gene Symbols IGFBP3
Host Species Rat
Immunogen Mouse recombinant protein of IGFBP-3
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3486
Test Specificity This antibody detects mouse IGFBP-3 in Western blotting. No cross activity to other mouse IGFBPs.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.