missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IA-2/PTPRN 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Lipid and Metabolism |
| Target | IA-2/PTPRN |
| Synonym | CA 512, EC 3.1.3.48, FLJ16131, IA2, IA-2, IA-2/PTP, ICA 512, ICA3, ICA512, islet cell antigen 2, Islet cell antigen 512, Islet cell autoantigen 3, protein tyrosine phosphatase, receptor type, N, protein tyrosine phosphatase-like N, PTP IA-2, receptor-type tyrosine-protein phosphatase-like N, R-PTP-N |
| Gene Symbol | PTPRN |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction