Learn More
R&D Systems™ I-BET 762
Description
I-BET 762 is a potent and high affinity BET bromodomain inhibitor (IC50 = 32.5 - 42.5 nM; Kd = 50.5 - 61.3 nM). Inhibits myeloma cell proliferation. Attenuates transcription of oncogenic MYC and suppresses key inflammatory genes. Exhibits efficacy in a range of oncology and immunoinflammatory models. Displays antiproliferative effect in vitro and in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (4S)-6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide |
| CAS | 1260907-17-2 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C22H22ClN5O2 |
| Purity | 0.98 |