missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Dimetridazole, TRC

Product Code. 17991365
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991365 5 mg 5mg
17963185 10 mg 10mg
17992475 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991365 Supplier Toronto Research Chemicals Supplier No. TRCH8252455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 936-05-0
Molecular Formula C5 H7 N3 O3
Molecular Weight (g/mol) 157.127
Synonym (1-Methyl-5-nitroimidazol-2-yl)methanol, Imidazole-2-methanol, 1-methyl-5-nitro- (7CI,8CI), 1-Methyl-5-nitro-1H-imidazole-2-methanol, (1-Methyl-5-nitro-1H-imidazol-2-yl)-methanol, 1-Methyl-2-hydroxymethyl-5-nitroimidazole, 1-Methyl-5-nitro-2-imidazolemethanol, 2-Hydroxymethyl-1-methyl-5-nitroimidazole, Hydroxydimetridazole
IUPAC Name (1-methyl-5-nitroimidazol-2-yl)methanol
SMILES Cn1c(CO)ncc1[N+](=O)[O-]

Tekniske data

Quantity 5 mg
InChI Formula InChI=1S/C5H7N3O3/c1-7-4(3-9)6-2-5(7)8(10)11/h2,9H,3H2,1H3
Formula Weight 157.049
Percent Purity >95%
Chemical Name or Material Dimetridazole-2-hydroxy
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.