accessibility menu, dialog, popup

UserName

CAS RN 936-05-0

CH3NOHNNOO

CAS RN 936-05-0

IUPAC Name: (1-methyl-2-nitro-1H-imidazol-5-yl)methanol
Synonyms: (3-methyl-2-nitroimidazol-4-yl)methanol
Molecular Weight (g/mol): 157.13
Molecular Formula: C5H7N3O3
InChi Key: WXSSUDRHAJTESR-UHFFFAOYSA-N
SMILES: CN1C(CO)=CN=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results