missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-5-nitroimidazol-2-yl)methanol, Mikromol™

Product Code. 17991026
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991026 100 mg 100mg
17902216 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991026 Supplier Mikromol Supplier No. MM1026.01
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1-Methyl-5-nitroimidazol-2-yl)methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimetridazole-2-hydroxy
CAS 936-05-0
Analyte or Component Names Dimetridazole-2-hydroxy
Shipping Condition Room Temperature
Molecular Formula C5 H7 N3 O3
Certifications/Compliance ISO 9001
Synonym (1-Methyl-5-nitroimidazol-2-yl)methanol, Imidazole-2-methanol, 1-methyl-5-nitro- (7CI,8CI), 1-Methyl-5-nitro-1H-imidazole-2-methanol, (1-Methyl-5-nitro-1H-imidazol-2-yl)-methanol, 1-Methyl-2-hydroxymethyl-5-nitroimidazole, 1-Methyl-5-nitro-2-imidazolemethanol, 2-Hydroxymethyl-1-methyl-5-nitroimidazole, Hydroxydimetridazole
InChI Formula InChI=1S/C5H7N3O3/c1-7-4(3-9)6-2-5(7)8(10)11/h2,9H,3H2,1H3
SMILES Cn1c(CO)ncc1[N+](=O)[O-]
IUPAC Name (1-methyl-5-nitroimidazol-2-yl)methanol
Molecular Weight (g/mol) 157.127
Quantity 100 mg
Formula Weight 157.049 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.