missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 40%, Supelco™

Product Code. 15883870
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15883870 2.5 L 2.5 L
15863870 500 mL 500mL
15873870 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15883870 Supplier Supelco Supplier No. 1003382500
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3

Specifications

Melting Point -44°C
Boiling Point 112°C/1013 hPa
Density 1.15 g/mL at 25°C,1.13 g/cm3 at 20°C
Quantity 2.5 L
Molecular Formula HF
MDL Number MFCD00011346
Synonym Hydrofluoric acid, Hydrofluoric acid solution, Fluoric acid, HF, Hydrofluoric acid, HF
Molecular Weight (g/mol) 20.01
Grade Ungraded
Chemical Name or Material Hydrofluoric Acid 40%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.