CAS RN 7664-39-3
Hydrofluoric Acid, 40%, Certified AR for Analysis, Fisher Chemical™
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 IUPAC Name: hydrogen fluoride SMILES: F
Hydrofluoric Acid, 48%, Certified AR for Analysis, Fisher Chemical™
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 SMILES: F
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 IUPAC Name: hydrogen fluoride SMILES: F
Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N IUPAC Name: hydrogen fluoride SMILES: F
Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 SMILES: F
Hydrofluoric Acid 47-51%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 IUPAC Name: hydrogen fluoride SMILES: F
Hydrofluoric acid, ACS, 48-51%, Thermo Scientific Chemicals
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 IUPAC Name: fluorane SMILES: F
Hydrofluoric Acid, Technical grade, Solstice
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228
Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%, Solstice
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228
Hydrofluoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥48%, Solstice
CAS: 7664-39-3 Molecular Formula: FH Molecular Weight (g/mol): 20.01 MDL Number: MFCD00011346 InChI Key: KRHYYFGTRYWZRS-UHFFFAOYSA-N Synonym: hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484 PubChem CID: 14917 ChEBI: CHEBI:29228 SMILES: F
Quality Control Standards 21 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Hydrofluoric Acid 48%, Supelco™
CAS: 7664-39-3