Learn More
Human TRIO knockout HEK-293T cell lysate, 1 Kit
Product Code. 32143864 Shop All Abcam Products
Product Code. 32143864 Supplier Abcam Supplier No. ab2592101Kit
 Restricted Item
Estimated Shipment: 13-11-2026
Human TRIO knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: TRIO knockout HEK-293T