Learn More
R&D Systems™ Human SLC7A6 (NP_001070253) VersaClone cDNA
Product Code. 18798458
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18798458 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18798458 Supplier R&D Systems™ Supplier No. RDC1375
  • ONLINE EXCLUSIVE PRICE
    341.00 EUR
    322.35 EUR / 10µg
    Save 18.65 EUR
    5% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Contains gene inserts that are fully sequenced and produced with high quality
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human SLC7A6 (NP_001070253) VersaClone cDNA clone contains the complete ORF for SLC7A6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC7A6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Gene SLC7A6
Accession Number NP_001070253
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.48
Insert Type 1561bp
Gene ID (Entrez) 9057
Gene Alias Y+L amino acid transporter 2, LAT-2, LAT3, solute carrier family 7 (amino acid transporter light chain, y+L system), member 6, y+LAT-2, Y+LAT2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
For Reasearch Use Only. Not for use in Humans.