Learn More
R&D Systems™ Human Pyruvate Carboxylase/PC (NP_071504.2) VersaClone cDNA
Specifications
Specifications
| Gene | Pyruvate Carboxylase |
| Accession Number | NP_071504.2 |
| Gene ID (Entrez) | 5091 |
| Gene Alias | EC 6.4.1, EC 6.4.1.1, PCBPyruvic carboxylase, pyruvate carboxylase, pyruvate carboxylase, mitochondrial |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |