Learn More
R&D Systems™ Human Proprotein Convertase 7 (NP_004707.2) VersaClone cDNA
Specifications
Specifications
| Gene | Proprotein Convertase 7/PCSK7 |
| Accession Number | NP_004707.2 |
| Gene ID (Entrez) | 9159 |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, EC 3.4.21.93, hPC8, LPC, LPCFLJ23503, Lymphoma proprotein convertase, PC7FLJ45329, PC8prohormone convertase PC7, PCSK7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, proprotein convertase subtilisin/kexin type 7, SPC7, SPC7proprotein convertase PC7, Subtilisin/kexin-like protease PC7 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |