Learn More
Human PPP1R13L (iASPP) knockout HeLa cell lysate, 1 Kit
Product Code. 32142831 Shop All Abcam Products
Product Code. 32142831 Supplier Abcam Supplier No. ab2585981Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Human PPP1R13L (iASPP) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: PPP1R13L (iASPP) knockout HeLa