Learn More
R&D Systems™ Human PON2 (NP_000296.2) VersaClone cDNA
Specifications
Specifications
| Gene | PON2 |
| Accession Number | NP_000296.2 |
| Gene ID (Entrez) | 5445 |
| Gene Alias | A-esterase 2, Aromatic esterase 2, EC 3.1.1.2, EC 3.1.1.81, paraoxonase 2, paraoxonase nirs, PON 2, Serum aryldialkylphosphatase 2, serum paraoxonase/arylesterase 2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |