Learn More
R&D Systems™ Human mGluR2 (NP_000830) VersaClone cDNA
Product Code. 18783519
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18783519 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18783519 Supplier R&D Systems™ Supplier No. RDC0989
Glutamate receptor, metabotropic 2
The Human mGluR2 (NP_000830) VersaClone cDNA clone contains the complete ORF for mGluR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans