Learn More
R&D Systems™ Human LRIG2 (NP_055628) VersaClone cDNA
Description
The Human LRIG2 (NP_055628) VersaClone cDNA clone contains the complete ORF for LRIG2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRIG2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
| Product Type | cDNA |
| Gene | hLRIG2 |
| Accession Number | NP_055628 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 3210bp |
| Gene ID (Entrez) | 9860 (Human); 269473 (Mouse); 310753 (Rat) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
For Research Use Only