Learn More
Human KIAA1191 knockout HeLa cell lysate, 1 Kit
Product Code. 32157455 Shop All Abcam Products
Product Code. 32157455 Supplier Abcam Supplier No. ab2632401Kit
 Restricted Item
Estimated Shipment: 30-11-2026
Human KIAA1191 knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: KIAA1191 knockout HeLa