Learn More
R&D Systems™ Human Histone Deacetylase 3/HDAC3(NP_003874) VersaClone cDNA
Specifications
Specifications
| Accession Number | NP_003874 |
| Gene ID (Entrez) | 8841 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |