missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FGF-23 (NP_065689) VersaClone cDNA
Description
- Also known as: ADHR; FGFN; HYPF; HFTC2; HPDR2; PHPTC
- FGF23 is a member of the fibroblast growth factor family of proteins, which possess broad mitogenic and cell survival activities and are involved in a variety of biological processes.
- It regulates phosphate homeostasis and transport in the kidney.
- The full-length, functional protein may be deactivated via cleavage into N-terminal and Cterminal chains.
- Mutation of this cleavage site causes autosomal dominant hypophosphatemic rickets (ADHR).
- Mutations in FGF23 are also associated with hyperphosphatemic familial tumoral calcinosis (HFTC).
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hFGF23 |
| Accession Number | NP_065689 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 769 bp |
| Gene ID (Entrez) | 8074 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?