Learn More
R&D Systems™ Human Connexin 26/GJB2 (NP_003995) VersaClone cDNA
Description
The Human Connexin 26/GJB2 (NP_003995) VersaClone cDNA clone contains the complete ORF for Connexin 26/GJB2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 26/GJB2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans