Learn More
R&D Systems™ Human Connexin 26/GJB2 (NP_003995) VersaClone cDNA
Product Code. 18704398
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18704398 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18704398 Supplier R&D Systems™ Supplier No. RDC1027
  • ONLINE EXCLUSIVE PRICE
    341.00 EUR
    322.35 EUR / 10µg
    Save 18.65 EUR
    5% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Gap junction protein, beta 2, 26kDa
The Human Connexin 26/GJB2 (NP_003995) VersaClone cDNA clone contains the complete ORF for Connexin 26/GJB2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 26/GJB2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans