Learn More
Human CDKN2B (p15 INK4b) knockout HeLa cell lysate, 1 Kit
Product Code. 32145488 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32145488 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32145488 Supplier Abcam Supplier No. ab2572671Kit
 Restricted Item
Estimated Shipment: 07-12-2026
Human CDKN2B (p15 INK4b) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: CDKN2B (p15 INK4b) knockout HeLa
Specifications
For Use With (Application) Sanger seq