Learn More
R&D Systems™ Human Activin C (NP_005529.1) VersaClone cDNA
Specifications
Specifications
| Gene | Activin C/Inhibin beta C |
| Accession Number | NP_005529.1 |
| Gene ID (Entrez) | 3626 |
| Gene Alias | inhibin beta C chain, IHBC, INHBC, inhibin, beta C |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |