missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Bovine, C. elegans, Canine, Chicken, Drosophila, Fish, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Plant, Rabbit, Rat, Sheep, Xenopus species samples. It is validated for Electron Microscopy, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | HSP70/HSPA1A |
| Applications | Electron Microscopy, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | N27F3-4 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | P08107 |
| Gene Alias | dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B |
| Gene Symbols | HSPA1A |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?