missing translation for 'onlineSavingsMsg'
Learn More

Histamine H2R Antibody, Novus Biologicals™

Product Code. 18214593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214593 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214593 Supplier Novus Biologicals Supplier No. NB600812
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody

Histamine H2R Polyclonal specifically detects Histamine H2R in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine H2R
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_071640.1
Gene Alias gastric receptor 1, Gastric receptor I, H2R, HH2R, histamine H2 receptor, histamine receptor H2
Gene Symbols HRH2
Host Species Goat
Immunogen Peptide with sequence C-QEEKPLKLQVWSGTE corresponding to C-Terminus according to NP_071640.1, NP_001124527.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3274
Test Specificity This antibody is expected to recognize both reported isoforms.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.