missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-3a-methyl-1,3-isobenzofurandione, TRC

Product Code. 30011763
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011763 10 mg 10mg
30009091 2.5 mg 2.5mg
30009388 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011763 Supplier Toronto Research Chemicals Supplier No. TRCH29378010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48122-14-1
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.1898
Synonym 1,3-Isobenzofurandione, hexahydro-3a-methyl-, 1,2-Cyclohexanedicarboxylic anhydride, 1-methyl- (6CI), 1-Methylhexahydrophthalic anhydride, 1-Methyl-1,2-Cyclohexanedicarboxylic anhydride
IUPAC Name 7a-methyl-4,5,6,7-tetrahydro-3aH-2-benzofuran-1,3-dione
SMILES CC12CCCCC1C(=O)OC2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H12O3/c1-9-5-3-2-4-6(9)7(10)12-8(9)11/h6H,2-5H2,1H3
Formula Weight 168.0786
Percent Purity >95%
Chemical Name or Material Hexahydro-3a-methyl-1,3-isobenzofurandione
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.