CAS RN 48122-14-1
CAS RN 48122-14-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexahydro-3a-methyl-1,3-isobenzofurandione, TRC
CAS: 48122-14-1 Molecular Formula: C9 H12 O3 Molecular Weight (g/mol): 168.1898 Synonym: 1,3-Isobenzofurandione, hexahydro-3a-methyl-,1,2-Cyclohexanedicarboxylic anhydride, 1-methyl- (6CI),1-Methylhexahydrophthalic anhydride,1-Methyl-1,2-Cyclohexanedicarboxylic anhydride IUPAC Name: 7a-methyl-4,5,6,7-tetrahydro-3aH-2-benzofuran-1,3-dione SMILES: CC12CCCCC1C(=O)OC2=O