missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 2 Antibody (T303), Alexa Fluor™ 700, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Herpes Simplex Virus 2 Monoclonal specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.
Specifications
Specifications
| Antigen | Herpes Simplex Virus 2 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | T303 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Gene Alias | Herpes Simplex Virus Type 2 |
| Host Species | Mouse |
| Immunogen | Exctract of Herpes Simplex Virus 2 infected VERO (green monkey kidney) cells. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?