missing translation for 'onlineSavingsMsg'
Learn More

Heptaethylene Glycol, TRC

Product Code. 30009785
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009785 500 mg 500mg
30004702 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009785 Supplier Toronto Research Chemicals Supplier No. TRCH279150500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5617-32-3
Molecular Formula C14H30O8
Molecular Weight (g/mol) 326.38
Synonym 3,6,9,12,15,18-Hexaoxaeicosane-1,20-diol;
IUPAC Name 2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES OCCOCCOCCOCCOCCOCCOCCO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H30O8/c15-1-3-17-5-7-19-9-11-21-13-14-22-12-10-20-8-6-18-4-2-16/h15-16H,1-14H2
Formula Weight 326.1941
Chemical Name or Material Heptaethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.