CAS RN 5617-32-3
CAS RN 5617-32-3
IUPAC Name:
3,6,9,12,15,18-hexaoxaicosane-1,20-diol
Synonyms:
heptaethylene glycol, peg330 peg330 heptaethylene glycol
Molecular Weight (g/mol):
326.39
Molecular Formula:
C14H30O8
InChi Key:
XPJRQAIZZQMSCM-UHFFFAOYSA-N
SMILES:
OCCOCCOCCOCCOCCOCCOCCO
1
–
1
of
1
results
1
Heptaethylene Glycol, TRC
CAS: 5617-32-3 Molecular Formula: C14H30O8 Molecular Weight (g/mol): 326.38 Synonym: 3,6,9,12,15,18-Hexaoxaeicosane-1,20-diol; IUPAC Name: 2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol SMILES: OCCOCCOCCOCCOCCOCCOCCO