Learn More
Thermo Scientific™ HEPES Lysis Buffer with Triton™ X-100
Product Code. 15496009
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496009 250mL 250mL
15406019 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15496009 Supplier Thermo Scientific™ Supplier No. J63860.AK
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
Content And Storage Ambient temperatures