missing translation for 'onlineSavingsMsg'
Learn More

Harmine, TRC

Product Code. 30011260
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011260 500 mg 500mg
30007623 1 g 1g
17920985 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011260 Supplier Toronto Research Chemicals Supplier No. TRCH105250500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13 H12 N2 O
Molecular Weight (g/mol) 212.25
Synonym 9H-Pyrido[3,4-b]indole, 7-methoxy-1-methyl-, 7-Methoxy-1-methyl-9H-pyrido[3,4-b]indole, Banisterin, Banisterine, Harmin, Harmine, Leucoharmine, Telepathin, Telepathine, Yagein, Yageine, 1-Methyl-7-methoxy-beta-carboline, 1-Methyl-7-methoxy-β-carboline
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COc1ccc2c(c1)[nH]c3c(C)nccc23

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H12N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-7,15H,1-2H3
Formula Weight 212.095
Chemical Name or Material Harmine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.