accessibility menu, dialog, popup

UserName

CAS RN 442-51-3

CH3ONHH3CN

CAS RN 442-51-3

IUPAC Name: 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
Synonyms: harmine yajeine leucoharmine banisterine (substance) banisterine telepathine yageine 6-methoxyharman
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: BXNJHAXVSOCGBA-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)