missing translation for 'onlineSavingsMsg'
Learn More

Guanylurea, TRC

Product Code. 17964015
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964015 10 mg 10mg
17932095 25 mg 25mg
17960515 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964015 Supplier Toronto Research Chemicals Supplier No. TRCG84449510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-83-3
Molecular Formula C2 H6 N4 O
Molecular Weight (g/mol) 102.1
Synonym Urea, (aminoiminomethyl)- (9CI), Urea, amidino- (6CI,7CI,8CI), N-(Aminoiminomethyl)urea, 1-Amidinourea, 1-Carbamoylguanidine, Amidinourea, Dicyandiamidine, Guanidine, (aminocarbonyl)-, Guanidinecarboxamide, Guanylurea, N-Amidinourea, N-Carbamoylguanidine, Urea, N-(aminoiminomethyl)-
IUPAC Name carbamimidoylurea
SMILES NC(=N)NC(=O)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C2H6N4O/c3-1(4)6-2(5)7/h(H6,3,4,5,6,7)
Formula Weight 102.0542
Chemical Name or Material Guanylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.