CAS RN 141-83-3
CAS RN 141-83-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Guanylurea, TRC
CAS: 141-83-3 Molecular Formula: C2 H6 N4 O Molecular Weight (g/mol): 102.1 Synonym: Urea, (aminoiminomethyl)- (9CI),Urea, amidino- (6CI,7CI,8CI),N-(Aminoiminomethyl)urea,1-Amidinourea,1-Carbamoylguanidine,Amidinourea,Dicyandiamidine,Guanidine, (aminocarbonyl)-,Guanidinecarboxamide,Guanylurea,N-Amidinourea,N-Carbamoylguanidine,Urea, N-(aminoiminomethyl)- IUPAC Name: carbamimidoylurea SMILES: NC(=N)NC(=O)N